Orders LinkContactUsLinkComponent
Volumetric flasks, class B
Volumetric flasks, class B
500 ml, 19/26, 265 mm
Catalog # 612-3004
Supplier:   KARTELL
Volumetric flasks, class B
Catalog # 612-3004
Supplier:  KARTELL
Volumetric flasks, class B
Volumetric flasks, class B
500 ml, 19/26, 265 mm
Price
23,50 €
Log in for availability
1 * 1 items
Product Details & Documents
PP, translucent, with PE stopper.
  • Narrow neck with no meniscus for easy reading
  • Calibrated gravimetrically at +20 °C
  • Autoclavable, chemically non absorbent, non wetting surface
Specifications
  • NS:
    19/26
  • Capacity:
    500 ml
  • Height:
    265 mm