Order Entry
Slovenia
Orders LinkContactUsLinkComponent
N,N-Dimethylformamide 99.5% for analysis

N,N-Dimethylformamide 99.5% for analysis

Glass bottle, 2,5 L
Catalog # 210580025.
Irritant
Health
Flammable
N,N-Dimethylformamide 99.5% for analysis
Catalog # 210580025.
CAS Number:  68-12-2
N,N-Dimethylformamide 99.5% for analysis
N,N-Dimethylformamide 99.5% for analysis

Glass bottle, 2,5 L

CAS Number:  68-12-2
Price
197,00 €
Log in for availability
1 * 2,5 L
Product Details & Documents
Synonyms:  Formic acid dimethylamide, Formdimethylamide, Dimethylformamide, DMF
  • CAS Number:68-12-2
  • Formula:HCON(CH₃)₂
  • MDL Number:MFCD00003284
  • Molecular Weight:73.09 g/mol
  • Merck Index:13,03269
  • Boiling Pt:153 °C (1013 hPa)
  • Melting Pt:–61 °C
  • Density:0.945 g/cm³
  • Flash Pt:58 °C (closed cup)
  • Storage Temperature:Ambient
  • EINECS:200-679-5
  • UN:2265
  • ADR:3,III
  • REACH:01-2119475605-32

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Water:
    ≤0.05 %
  • GC:
    ≥99.5 %
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤20 ppm
  • Acidity/alkalinity:
    ≤0.0005 meq/g
  • Calcium (Ca):
    ≤1 ppm
  • Copper (Cu):
    ≤0.5 ppm
  • Iron (Fe):
    ≤1 ppm
  • Lead (Pb):
    ≤0.5 ppm
  • Magnesium (Mg):
    ≤0.5 ppm
  • Potassium (K):
    ≤0.5 ppm
  • Sodium (Na):
    ≤2 ppm
  • Zinc (Zn):
    ≤0.5 ppm
  • Total phosphorus (P):
    ≤0.5 ppm
  • Total silicon (Si):
    ≤0.05 ppm
  • Total sulfur (S):
    ≤1 ppm

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 L

Specifications