Order Entry
Slovenia
Orders LinkContactUsLinkComponent
2-Chlorophenol 98+%

2-Chlorophenol 98+%

Glass bottle for solids, 500 g
Catalog # ACRO180995000
Irritant
Environment
2-Chlorophenol 98+%
Catalog # ACRO180995000
CAS Number:  95-57-8
2-Chlorophenol 98+%
2-Chlorophenol 98+%

Glass bottle for solids, 500 g

CAS Number:  95-57-8
Price
57,20 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  1-Chloro-2-hydroxybenzene
  • CAS Number:95-57-8
  • Formula:C₆H₅ClO
  • MDL Number:MFCD00002159
  • Molecular Weight:128.56 g/mol
  • Merck Index:12,02173
  • Boiling Pt:173…175 °C (760 mmHg)
  • Melting Pt:8 °C
  • Density:1.262 g/cm³
  • Flash Pt:63 °C
  • Storage Temperature:Ambient
  • EINECS:202-433-2
  • UN:2021
  • ADR:6.1,III

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear light yellow to brown Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5580 to 1.5600 (20°C, 589 nm)
  • GC:
    ≥98.0 %

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.