Thermometer probes
Flexible wire type K air probe, with sub-miniature connector, AK31M, −100…+250 °C, - mm
COMAAK31M
: COMARK
- Range:−100…+250 °C
- Description:Flexible wire type K air probe, with sub-miniature connector, AK31M
- Ø×L:- mm