Order Entry
Slovenia
Orders LinkContactUsLinkComponent
 

3-Chlorophenol 99%

Glass bottle for solids, 100 g
Catalog # ACRO109811000
Irritant
Environment
3-Chlorophenol 99%
Catalog # ACRO109811000
CAS Number:  108-43-0
3-Chlorophenol 99%
3-Chlorophenol 99%

Glass bottle for solids, 100 g

CAS Number:  108-43-0
Price
86,50 €
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:108-43-0
  • Formula:C₆H₅ClO
  • MDL Number:MFCD00002256
  • Molecular Weight:128.56 g/mol
  • Merck Index:13,02173
  • Boiling Pt:214 °C (760 mmHg)
  • Melting Pt:33…35 °C
  • Density:1.218 g/cm³
  • Flash Pt:97 °C
  • Storage Temperature:Ambient
  • EINECS:203-582-6
  • UN:2020
  • ADR:6.1,III
Specifications

Specification Test Results

  • Appearance:
    Clear yellow to brown Liquid after melting
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.5650 to 1.5670 (20°C, 589 nm)
  • GC:
    ≥98.5 %
  • Iron (Fe):
    ≤50 ppm
  • Magnesium (Mg):
    ≤100 ppm
  • Potassium (K):
    ≤50 ppm
  • Sodium (Na):
    ≤100 ppm
  • Silicon dioxide (SiO2):
    ≤1000 ppm

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications