2-Chlorocyclohexanone 95% stabilised
Plastic bottle, 100 g
Supplier: Thermo Fisher Scientific
2-Chlorocyclohexanone 95% stabilised
Catalog # ACRO108961000
Supplier: Thermo Fisher Scientific
CAS Number: 822-87-7
Jump to:
Product Details & Documents
- CAS Number:822-87-7
- Formula:C₆H₉ClO
- MDL Number:MFCD00001626
- Molecular Weight:132.59 g/mol
- Boiling Pt:82…83 °C (10 mmHg)
- Melting Pt:23 °C
- Density:1.16 g/cm³
- Flash Pt:82 °C
- Storage Temperature:Refrigerator
- EINECS:212-505-5
- ADR:9,
Documents
ACRO10896
SDS
Specifications
Specification Test Results
- Appearance:Clear brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4830 to 1.4850 (20°C, 589 nm)
- GC:≥94 %
- Stabilizer:0.2 % CaCO3/MgO (1:1)
Specifications
- Pack type:Plastic bottle
- Pk:100 g



