Orders LinkContactUsLinkComponent
Containers with screw cap, multipurpose
Containers with screw cap, multipurpose
500 ml, 104 mm, 100 mm, Transparent
Catalog # BURK6202-0500
Supplier:   BURKLE
Containers with screw cap, multipurpose
Catalog # BURK6202-0500
Supplier:  BURKLE
Containers with screw cap, multipurpose
Containers with screw cap, multipurpose
500 ml, 104 mm, 100 mm, Transparent
Price
6,80 €
Log in for availability
1 * 1 items
Product Details & Documents
PP, translucent, with PE screw lid.
  • Suitable for use with foodstuffs
  • No crevices
  • Very good chemical and temperature resistance
Contents can be recognised easily due to the transparent sides. Emptying without residues.
Specifications
  • Ext.Ø:
    104 mm
  • Capacity:
    500 ml
  • Colour:
    Transparent
  • Height:
    100 mm