Sample dippers
Sampling dipper with reinforced rim and spout, PTFE/steel, 155 mm, 105 mm, 1000 ml
Catalog # BRND90462
Supplier: Brand
Jump to:
Product Details & Documents
Sampling dipper with reinforced rim and spout.
- PTFE handle with steel core
Optional extension rod for increased handle length.
Specifications
- Ø:105 mm
- Capacity:1000 ml
- Description:Sampling dipper with reinforced rim and spout
- Length:155 mm
- Material:PTFE/steel
- Cat. No.:BRND90462
Accessories
Product Information
- Catalog #AvailabilityPrice
- Extension rod for sampling device, 600 mm