Orders LinkContactUsLinkComponent
Buckets
Buckets
Transparent, 12 L, 300 mm
Catalog # BRND72376
Supplier:   Brand
Buckets
Catalog # BRND72376
Supplier:  Brand
Buckets
Buckets
Transparent, 12 L, 300 mm
Price
35,30 €
Log in for availability
1 * 1 items
Product Details & Documents
PP, translucent.
  • Graduated
  • Very good thermal resistance
  • Very good chemical resistance
Universal multipurpose bucket with spout, reinforced rim and handle.
Specifications
  • Capacity:
    12 L
  • Colour:
    Transparent
  • Height:
    300 mm