Basic fuchsin 88%, pure
Glass bottle for solids, 100 g
: CI 42510, Fuchsin basic, Magenta O, Basic-fuchsin, Rosaniline hydrochloride, 4-((4-Aminophenyl)(4-iminocyclohexa-2,5-dien-1-ylidene)methyl)-2-methylaniline hydrochloride, Fuchsine base monohydrochloride, Basic fuchsin hydrochloride, (4-(4-Aminophenyl)(4-iminocyclohexa-2,5-dienylidene)methyl)-2-methylaniline hydrochloride, Diamondfuchsin, Basic fuchsin, Basic Red 9, Pararosaniline
- :632-99-5
- :C₂₀H₂₀ClN₃
- :MFCD00012569
- :337.85 g/mol
- :12,05686
- :250 °C (dec.)
- :1,22 g/cm³ (20 °C)
- :200 °C
- :Ambient
- :211-189-6
Documents
- Appearance:Green Powder
- Infrared spectrum:Conforms
- Loss on drying:≤10 % (105°C)
- Dye content:≥83 % (on dry substance)
- UV Lambda max:549 to 555 nm (in 50:50 IMS:water)
- Absorption Ratio P-15/P+15:1.1 to 1.4
- Specific absorbance A (1%/1cm):≥1800 (at lambda max.)
- Pack type:Glass bottle for solids
- Pk:100 g



