Orders LinkContactUsLinkComponent
Dissolved oxygen bottles
Oxygen bottles acc. to Winkler, with stopper, 100 - 150 ml, 14/23
  528-1303
 :   Brand
Dissolved oxygen bottles
  528-1303
 :  Brand
Dissolved oxygen bottles
Dissolved oxygen bottles
Oxygen bottles acc. to Winkler, with stopper, 100 - 150 ml, 14/23
28,60 €
1 * 1 items
Soda-lime glass.
  • Solid, obliquely cut standard-ground glass stopper can be secured with a fastening clip
  • Each bottle is calibrated with its own stopper; hence, stoppers and bottles are not interchangeable
  • Each bottle and its stopper is marked with a unique matching identification number
For the determination of oxygen dissolved water. The measured volume is indicated to ± 0,01 ml. White labelling area.
  • Capacity:
    100 - 150 ml
  • Description:
    Oxygen bottles acc. to Winkler, with stopper
  • NS stopper:
    14/23
Accessories
  •  
    Description
  • 528-1311
    Fastening clip for dissolved oxygen bottles, winkler pattern with stopper NS 19/26
    1 * 1 items - 7,00 €
  •  
    Description
  • BRND386138
    Fastening clip for dissolved oxygen bottles, winkler pattern with stopper NS 14/23
    1 * 1 items - 6,60 €