Digitonin
Glass bottle, 500 mg
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Synonyms: Digitonin
- CAS Number:11024-24-1
- Formula:C₅₆H₉₂O₂₉
- MDL Number:MFCD00077729
- Molecular Weight:1229.312 g/mol
- Melting Pt:240 °C
- Storage Temperature:Ambient
- EINECS:234-255-6
- UN:3462
- ADR:6.1,III
Documents
ACRO40756
SDS
Specifications
Specification Test Results
- Appearance:White to yellow Powder
- Infrared spectrum:Conforms
- Loss on drying:≤6 % (1 g, 105°C)
- Appearance of solution:(2.5% in warm EtOH) clear colorless to faint yellow
- Residue after ignition:≤0.3 %
- Specific optical rotation:-45° to -51° (20°C, 589 nm) (c=10, 75% acetic acid)
Specifications
- Pack type:Glass bottle
- Pk:500 mg