Orders LinkContactUsLinkComponent
Measuring scoops
Measuring scoops
250 ml, 260 mm
Catalog # 231-0338
Supplier:   Brand
Measuring scoops
Catalog # 231-0338
Supplier:  Brand
Measuring scoops
Measuring scoops
250 ml, 260 mm
Price
25,80 €
Log in for availability
1 * 6 items
Product Details & Documents
PP, white. With flat, bevelled end and reinforced handle.
  • Excellent chemical resistance, unbreakable
  • The capacity of the scoop is stamped into the handle for easy reading
Specifications
  • Capacity:
    250 ml
  • Length:
    260 mm