Orders LinkContactUsLinkComponent
Measuring scoops
Measuring scoops
1000 ml, 400 mm
Catalog # 231-0114
Supplier:   Brand
Measuring scoops
Catalog # 231-0114
Supplier:  Brand
Measuring scoops
Measuring scoops
1000 ml, 400 mm
Price
58,20 €
Log in for availability
1 * 6 items
Product Details & Documents
HDPE, natural coloured. With flat, bevelled end.
  • Good chemical resistance
  • Good thermal resistance
  • Robust, unbreakable construction
Specifications
  • Capacity:
    1000 ml
  • Length:
    400 mm
  • Seq:
    03