Orders LinkContactUsLinkComponent
Bossheads for frames, 90°
Bossheads for frames, 90°
13 mm, Stainless steel 18/10, anti-magnetic
Catalog # 241-7185
Supplier:   BOCHEM
Bossheads for frames, 90°
Catalog # 241-7185
Supplier:  BOCHEM
Bossheads for frames, 90°
Bossheads for frames, 90°
13 mm, Stainless steel 18/10, anti-magnetic
Price
45,20 €
Log in for availability
1 * 1 items
Product Details & Documents
For rods up to 13 mm.
  • Angle: 90°
  • Heart screws
Specifications
  • Spanning width:
    13 mm
  • Material:
    Stainless steel 18/10, anti-magnetic