Orders LinkContactUsLinkComponent
Laboratory spoons
280 mm, 85×70 mm
Catalog # 231-1215
Supplier:   BOCHEM
Laboratory spoons
Catalog # 231-1215
Supplier:  BOCHEM
Laboratory spoons
Laboratory spoons
280 mm, 85×70 mm
Price
17,90 €
Log in for availability
1 * 1 items
Product Details & Documents
Stainless steel 18/10, anti-magnetic. Manual scoops for collecting or measuring powders, pastes or granulates.
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Robust design
Specifications
  • Length:
    280 mm
  • Spoon size:
    85×70 mm