Laboratory spoons
195 mm, 60×45 mm
231-1214
: BOCHEM
Stainless steel 18/10, anti-magnetic. Manual scoops for collecting or measuring powders, pastes or granulates.
- Excellent thermal resistance, autoclavable
- Good chemical resistance
- Robust design
- Length:195 mm
- Spoon size:60×45 mm
- Cat. No.:231-1214