Orders LinkContactUsLinkComponent
Laboratory spoons
Laboratory spoons
180 mm, 55×38 mm
Catalog # 231-1213
Supplier:   BOCHEM
Laboratory spoons
Catalog # 231-1213
Supplier:  BOCHEM
Laboratory spoons
Laboratory spoons
180 mm, 55×38 mm
Price
4,50 €
Log in for availability
1 * 1 items
Product Details & Documents
Stainless steel 18/10, anti-magnetic. Manual scoops for collecting or measuring powders, pastes or granulates.
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Robust design
Specifications
  • Length:
    180 mm
  • Spoon size:
    55×38 mm
  • Cat. No.:
    231-1213