Orders LinkContactUsLinkComponent
Pharmacist spoons
Pharmacist spoons
Spoon in scoop shape, 170 mm, 40×10 mm
Catalog # BOCH3360
Supplier:   BOCHEM
Pharmacist spoons
Catalog # BOCH3360
Supplier:  BOCHEM
Pharmacist spoons
Pharmacist spoons
Spoon in scoop shape, 170 mm, 40×10 mm
Price
2,60 €
Log in for availability
1 * 1 items
Product Details & Documents
Stainless steel 18/10, anti-magnetic. Multipurpose spoons for a wide range of uses.
  • Narrow shape with extra long, flat handle
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Quick and easy to clean
Specifications
  • Description:
    Spoon in scoop shape
  • Length:
    170 mm
  • Spoon size:
    40×10 mm