Orders LinkContactUsLinkComponent
Spoon spatulas
Spoon spatulas
185 mm, 10×5 mm
Catalog # 231-1325
Supplier:   BOCHEM
Spoon spatulas
Catalog # 231-1325
Supplier:  BOCHEM
Spoon spatulas
Spoon spatulas
185 mm, 10×5 mm
Price
2,10 €
Log in for availability
1 * 1 items
Product Details & Documents
Stainless steel. Universal spoon spatula designed for a wide range of uses.
  • One end is shaped like a spoon and the other end like a spatula
  • With rounded handle
  • High thermal resistance, autoclavable
  • Excellent resistance to chemicals
  • Quick and easy to clean
Documents
Specifications
  • Length:
    185 mm
  • Spoon size:
    10×5 mm