Orders LinkContactUsLinkComponent
Forceps, precision
Forceps, precision
Straight, 105 mm
Catalog # 232-0007
Supplier:   BOCHEM
Forceps, precision
Catalog # 232-0007
Supplier:  BOCHEM
Forceps, precision
Forceps, precision
Straight, 105 mm
Price
7,50 €
Log in for availability
1 * 1 items
Product Details & Documents
Stainless steel ¹⁸/₁₀, anti-magnetic. Extremely precise forceps designed for a wide variety of uses in the laboratory, production and more.
  • Extremely sharp, flat
  • Non serrated points
Specifications
  • Type:
    Straight
  • Length:
    105 mm