Orders LinkContactUsLinkComponent
Inoculating loops and needles
Inoculating loops and needles
Loop, Ø 4,0 mm
Catalog # 631-1403
Supplier:   BOCHEM
Inoculating loops and needles
Catalog # 631-1403
Supplier:  BOCHEM
Inoculating loops and needles
Inoculating loops and needles
Loop, Ø 4,0 mm
Price
1,70 €
Log in for availability
1 * 1 items
Product Details & Documents
Stainless steel 18/10. Length 100 mm.
Specifications
  • Description:
    Loop, Ø 4,0 mm