Forceps
Straight, sharp, 115 mm
Catalog # BOCH1011
Supplier: BOCHEM
Jump to:
Product Details & Documents
Precision forceps made from stainless steel 18/10.
Documents
Certificates
Search by Lot NumberSpecifications
- Type:Straight, sharp
- Length:115 mm
- Seq:02