Orders LinkContactUsLinkComponent
Cell scrapers
Cell scrapers
Cell scraper, Medium, replacement blade, Stainless steel, Blade width (mm): 31,75 mm
Catalog # BELC7731-22005
Supplier:   BELLCO GLASS
Cell scrapers
Catalog # BELC7731-22005
Supplier:  BELLCO GLASS
Cell scrapers
Cell scrapers
Cell scraper, Medium, replacement blade, Stainless steel, Blade width (mm): 31,75 mm
Price
377,00 €
Log in for availability
1 * 10 items
Product Details & Documents
Stainless steel blade holder mounted on plastic covered stainless steel handle.
Specifications
  • Description:
    Cell scraper, Medium, replacement blade, Stainless steel, Blade width (mm): 31,75 mm