Orders LinkContactUsLinkComponent
Overhead stirrer tools, paddle stirrers
Overhead stirrer tools, paddle stirrers
Paddle stirrer, 70 mm, 8 mm, 550 mm, 800 min-1
Catalog # 441-2942
Supplier:   IKA
Overhead stirrer tools, paddle stirrers
Catalog # 441-2942
Supplier:  IKA
Overhead stirrer tools, paddle stirrers
Overhead stirrer tools, paddle stirrers
Paddle stirrer, 70 mm, 8 mm, 550 mm, 800 min-1
Price
224,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Made from stainless steel. Gentle stirring with minimal turbulence.
  • Ideal for stirring liquids at low to medium speeds
  • Creates tangential flow
Specifications
  • Description:
    Paddle stirrer
  • Seq:
    01
  • Max. revolutions:
    800 min-1
  • Shaft Ø:
    8 mm
  • Shaft length:
    550 mm
  • Stirrer Ø:
    70 mm