Orders LinkContactUsLinkComponent
Transfer capillaries
Transfer capillaries
100 mm, 2,35 mm, 1,55 mm
Catalog # HIRS9202310
Supplier:   HIRSCHMANN
Transfer capillaries
Catalog # HIRS9202310
Supplier:  HIRSCHMANN
Transfer capillaries
Transfer capillaries
100 mm, 2,35 mm, 1,55 mm
Price
123,00 €
Log in for availability
1 * 1.000 items
Product Details & Documents
Clear glass, both end open.
Specifications
  • Ext.Ø:
    2,35 mm
  • Ø int.:
    1,55 mm
  • Length:
    100 mm
  • Seq:
    32