Orders LinkContactUsLinkComponent
Transfer capillaries
100 mm, 1,00 mm, 0,80
Catalog # 612-2804
Supplier:   HIRSCHMANN
Transfer capillaries
Catalog # 612-2804
Supplier:  HIRSCHMANN
Transfer capillaries
Transfer capillaries
100 mm, 1,00 mm, 0,80
Price
101,00 €
Log in for availability
1 * 1.000 items
Product Details & Documents
Clear glass, both end open.
Specifications
  • Ext.Ø:
    1,00 mm
  • Ø int.:
    0,80
  • Length:
    100 mm
  • Seq:
    05