Orders LinkContactUsLinkComponent
Automatic burettes, Dr. Schilling pattern, Schellbach
Automatic burettes, Dr. Schilling pattern, Schellbach
PP base for 1000 ml bottles for Schilling burettes
Catalog # 612-2803
Supplier:   HIRSCHMANN
Automatic burettes, Dr. Schilling pattern, Schellbach
Catalog # 612-2803
Supplier:  HIRSCHMANN
Automatic burettes, Dr. Schilling pattern, Schellbach
Automatic burettes, Dr. Schilling pattern, Schellbach
PP base for 1000 ml bottles for Schilling burettes
Price
8,60 €
Log in for availability
1 * 1 items
Product Details & Documents
Borosilicate 3.3 glass, silicone tubing, PVC fitting, PE bottle and PP base.
  • Schellbach stripe, blue graduations
  • Automatic zeroing, squeezing bottle allows rapid filling, micrometre screw allows fine titration
With bottle: 500 ml for 5 and 10 ml; 1000 ml for 15, 25 and 50 ml.
Specifications
  • Description:
    PP base for 1000 ml bottles for Schilling burettes
  • Seq:
    07
Accessories
Product Information
  • Catalog #
    Capacity
    Division
    Tolerance
    Availability
    Price
  • 612-2798
    10 ml
    0,05 ml
    0,03 ± ml
    Log in for availability
    1 * 1 items - 47,10 €
Product Information
  • Catalog #
    Capacity
    Division
    Tolerance
    Availability
    Price
  • 612-2797
    5 ml
    0,05 ml
    0,03 ± ml
    Log in for availability
    1 * 1 items - 48,50 €
Product Information
  • Catalog #
    Capacity
    Division
    Tolerance
    Availability
    Price
  • 612-2801
    50 ml
    0,1 ml
    0,075 ± ml
    Log in for availability
    1 * 1 items - 54,30 €
Product Information
  • Catalog #
    Capacity
    Division
    Tolerance
    Availability
    Price
  • 612-2799
    15 ml
    0,1 ml
    0,045 ± ml
    Log in for availability
    1 * 1 items - 47,10 €
Product Information
  • Catalog #
    Capacity
    Division
    Tolerance
    Availability
    Price
  • 612-2800
    25 ml
    0,1 ml
    0,045 ± ml
    Log in for availability
    1 * 1 items - 49,90 €
Product Information
  • Catalog #
    Availability
    Price
  • Fittings for Dr Schilling burette 5 and 10 ml
    Fittings for Dr Schilling burette 5 and 10 ml
    HIRS8900710
    Log in for availability
    1 * 1 items - 22,60 €
Product Information
  • Catalog #
    Availability
    Price
  • Fittings for Dr Schilling burette 15-50 ml
    Fittings for Dr Schilling burette 15-50 ml
    HIRS8900750
    Log in for availability
    1 * 1 items - 22,60 €