Automatic burettes, Dr. Schilling pattern, Schellbach
PP base for 500 ml bottles for Schilling burettes
Catalog # 612-2802
Supplier: HIRSCHMANN
Jump to:
Product Details & Documents
Borosilicate 3.3 glass, silicone tubing, PVC fitting, PE bottle and PP base.
- Schellbach stripe, blue graduations
- Automatic zeroing, squeezing bottle allows rapid filling, micrometre screw allows fine titration
With bottle: 500 ml for 5 and 10 ml; 1000 ml for 15, 25 and 50 ml.
Documents
Specifications
- Description:PP base for 500 ml bottles for Schilling burettes
- Seq:06
Accessories
Product Information
- Catalog #CapacityDivisionToleranceAvailabilityPrice
Product Information
- Catalog #CapacityDivisionToleranceAvailabilityPrice
Product Information
- Catalog #CapacityDivisionToleranceAvailabilityPrice
Product Information
- Catalog #CapacityDivisionToleranceAvailabilityPrice
Product Information
- Catalog #CapacityDivisionToleranceAvailabilityPrice
Product Information
- Catalog #AvailabilityPrice
- Fittings for Dr Schilling burette 5 and 10 mlFittings for Dr Schilling burette 5 and 10 mlHIRS8900710Log in for availability1 * 1 items - 22,60 €
Product Information
- Catalog #AvailabilityPrice
- Fittings for Dr Schilling burette 15-50 mlFittings for Dr Schilling burette 15-50 mlHIRS8900750Log in for availability1 * 1 items - 22,60 €