Orders LinkContactUsLinkComponent
Volumetric flasks, class B
Volumetric flasks, class B
500 ml, 19/26, 270 mm
Catalog # HIRS7550190
Supplier:   HIRSCHMANN
Volumetric flasks, class B
Catalog # HIRS7550190
Supplier:  HIRSCHMANN
Volumetric flasks, class B
Volumetric flasks, class B
500 ml, 19/26, 270 mm
Price
40,30 €
Log in for availability
1 * 2 items
Product Details & Documents
PP, translucent, with PP NS-stopper.
Specifications
  • NS:
    19/26
  • Capacity:
    500 ml
  • Height:
    270 mm