Volumetric flasks, class A, blue markings
5000 ml, 34/35, 470 ± 5 mm, 1,200 ± ml
Catalog # 612-3503
Supplier: HIRSCHMANN
Jump to:
Product Details & Documents
DURAN®, borosilicate glass, without stopper, conformity certified (DE-M marking).
- Blue enamelled indelible markings resistant to acids and alkalis
- Certificate of conformity with dated batch identification available via website: (www.hirschmannlab.de)
*Wide neck
Certifications: DIN EN ISO 1042
Documents
Specifications
- NS:34/35
- Capacity:5000 ml
- Height:470 ± 5 mm
- Seq:33
- Tolerance:1,200 ± ml
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



