Orders LinkContactUsLinkComponent
Analogue stopwatches, split-second addition timer
Analogue stopwatches, split-second addition timer
Analogue stopwatch, lever, 7 jewels, 1/5 s + 1/100 min, 30 min, 130 g, 55 mm
Catalog # 609-0146
Supplier:   HANHART
Analogue stopwatches, split-second addition timer
Catalog # 609-0146
Supplier:  HANHART
Analogue stopwatches, split-second addition timer
Analogue stopwatches, split-second addition timer
Analogue stopwatch, lever, 7 jewels, 1/5 s + 1/100 min, 30 min, 130 g, 55 mm
Price
714,00 €
Log in for availability
1 * 1 items
Product Details & Documents
Addition timer for time measurement with interrupt option, also drag indicator for split-time measurement.
  • Chrome plated brass casing, diamond cut
  • Lever, 7 jewels
  • Shock resistant, additional dust and water protection
Specifications
  • Ø:
    55 mm
  • Description:
    Analogue stopwatch, lever, 7 jewels, 1/5 s + 1/100 min, 30 min, 130 g