Orders LinkContactUsLinkComponent
Spoons
Spoons
140 mm, 45×30 mm
Catalog # 231-0270
Supplier:   HAMMACHER KARL
Spoons
Catalog # 231-0270
Supplier:  HAMMACHER KARL
Spoons
Spoons
140 mm, 45×30 mm
Price
17,00 €
Log in for availability
1 * 1 items
Product Details & Documents
WIRONIT steel, anti-magnetic.
Specifications
  • Length:
    140 mm
  • Spoon size:
    45×30 mm