BUFFER SET PH 4,01/7,00/9,21 1 * 3 items
Catalog # 662-9248
Supplier: HAMILTON BONADUZ
Frequently Bought Together
VWR®, Erlenmeyer Flasks
Supplier: VWR Collection
VWR® Facial Tissues
Supplier: VWR Collection
Sterican® Single-Use Needles for Special Applications
Supplier: B. Braun
Acetic acid ≥99.8% (by HPLC), HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
VWR®, Petri Dish, PS
Supplier: VWR Collection