Volumetric flasks, class A, blue markings
50 ml, 12/21, 0,06 ± ml
Catalog # 612-0543
Supplier: Poulten & Graf
Jump to:
Product Details & Documents
Borosilicate glass 3.3 with PP stopper.
- Calibrated to contain (TC, In)
- With ground ring mark
- Conformity approved
Certifications: DIN EN ISO 1042
Specifications
- NS:12/21
- Capacity:50 ml
- Seq:05
- Tolerance:0,06 ± ml