Orders LinkContactUsLinkComponent
Test sieves, 200×50 mm
200 mm, 50 mm, 4,00 mm
Catalog # ENDE200SAW4.00
Supplier:   ENDECOTTS
Test sieves, 200×50 mm
Catalog # ENDE200SAW4.00
Supplier:  ENDECOTTS
Test sieves, 200×50 mm
Test sieves, 200×50 mm
200 mm, 50 mm, 4,00 mm
Price
306,00 €
Log in for availability
1 * 1 items
Product Details & Documents
These test sieves are made from stainless steel.
 : According to ASTM E 11-95
 : Each sieve is delivered with an individual quality certificate.
Specifications
  • Ø:
    200 mm
  • Mesh size:
    4,00 mm
  • Height:
    50 mm