Orders LinkContactUsLinkComponent
Test sieves, 200×50 mm
200 mm, 50 mm, 1,40 mm
  ENDE200SAW1.40
 :   ENDECOTTS
Test sieves, 200×50 mm
  ENDE200SAW1.40
 :  ENDECOTTS
Test sieves, 200×50 mm
Test sieves, 200×50 mm
200 mm, 50 mm, 1,40 mm
241,00 €
1 * 1 items
These test sieves are made from stainless steel.
 : According to ASTM E 11-95
 : Each sieve is delivered with an individual quality certificate.
  • Ø:
    200 mm
  • Mesh size:
    1,40 mm
  • Height:
    50 mm