ContactUsLinkComponent
Tetracon 700-1,5,universal 4 electrode conductivity cell especially for waste water treatment plants, for use with ecoline and quad roline monitorswith integrated temperature sensor, 1.5 m cable and watertight plug (ip68) 1 * 1 items
Catalog # WTWO302314
Supplier:  WTW ONLINE
Tetracon 700-1,5,universal 4 electrode conductivity cell especially for waste water treatment plants, for use with ecoline and quad roline monitorswith integrated temperature sensor, 1.5 m cable and watertight plug (ip68) 1 * 1 items
Catalog # WTWO302314
Supplier:  WTW ONLINE
Frequently Bought Together