ContactUsLinkComponent
Ki/ph 170,junction box with integrated pre-amplifier for high-impedance connection of a ph electrode and a temperature sensor (ntc, pt 100, pt 1000) 1 * 1 items
Catalog # WTWO108596
Supplier:  WTW ONLINE
Ki/ph 170,junction box with integrated pre-amplifier for high-impedance connection of a ph electrode and a temperature sensor (ntc, pt 100, pt 1000) 1 * 1 items
Catalog # WTWO108596
Supplier:  WTW ONLINE
Frequently Bought Together