ContactUsLinkComponent
As 7-s-1,5,connecting cable for high-impedance connection of ph combination electrodes with s8 plug head to junction boxes (e.g. ki/ ph 170)cable length 1.5 m 1 * 1 items
Catalog # WTWO108511
Supplier:  WTW ONLINE
As 7-s-1,5,connecting cable for high-impedance connection of ph combination electrodes with s8 plug head to junction boxes (e.g. ki/ ph 170)cable length 1.5 m 1 * 1 items
Catalog # WTWO108511
Supplier:  WTW ONLINE
Frequently Bought Together