Temperature probes
Temperature probe, with 1 m cable and water proof plug, TFK 150
Catalog # 663-1821
Supplier: WTW
Jump to:
Product Details & Documents
Specifications
- Description:Temperature probe, with 1 m cable and water proof plug
- Type:TFK 150
- Application:Automatic temperature compensation
Accessories
Product Information
- Catalog #AvailabilityPrice
- Holding clip for SenTix pH electrodes and temperature sensor TFK 325/HCHolding clip for SenTix pH electrodes and temperature sensor TFK 325/HCWTWA109821Log in for availability1 * 1 items - 110,00 €