EasyPlus™ Titration Electrodes
EM43-BNC Sensor Redox Ipol, ±2000 mV, Easy Pro, Easy Ox titrators
Catalog # METT30043105
New Product
Supplier: Mettler - Toledo
Jump to:
Product Details & Documents
Electrode for use with Mettler Toledo® range of EasyPlus™ titrators only.
Electrode only.
Documents
Specifications
- For:Easy Pro, Easy Ox titrators
- Description:EM43-BNC Sensor Redox Ipol
- Seq:4
- Range:±2000 mV