Orders LinkContactUsLinkComponent
Chlorophenol red
Chlorophenol red
Bottle
Catalog # APOSBI4558-5G
Irritant
New Product
Supplier:   Apollo Scientific
Chlorophenol red
Catalog # APOSBI4558-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  4430-20-0
Chlorophenol red
Bottle
CAS Number:  4430-20-0
Price
39,10 €
Log in for availability
1 * 5 g
Product Details & Documents
Usage: Biochemical/ life science applications.
Synonyms:  3,3-Bis(3-chloro-4-hydroxyphenyl)-3H-benzo[c][1,2]oxathiole 1,1-dioxide, 4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2-chlorophenol] S,S-dioxide
  • CAS Number:4430-20-0
  • Formula:C₁₉H₁₂Cl₂O₅S
  • MDL Number:MFCD00005877
  • Molecular Weight:423,273 g/mol
  • EINECS:224-619-2
Specifications
Specifications
  • Pack type:
    Bottle