Orders LinkContactUsLinkComponent
Estrone ≥98%
Estrone ≥98%
Bottle
Catalog # APOSBUP18515-500MG
New Product
Supplier:   Apollo Scientific
Estrone ≥98%
Catalog # APOSBUP18515-500MG
Supplier:  Apollo Scientific
New Product
CAS Number:  53-16-7
Estrone ≥98%
Bottle
CAS Number:  53-16-7
Price
59,40 €
Log in for availability
1 * 500 mg
Product Details & Documents
Usage: Biochemical/life science applications.
  • CAS Number:53-16-7
  • Formula:C₁₈H₂₂O₂
  • Molecular Weight:270,37 g/mol
Specifications
Specifications
  • Pack type:
    Bottle