Orders LinkContactUsLinkComponent
Hydroxyectoin 95%
Bottle
Catalog # APOSOR1025890-5G
New Product
Supplier:   Apollo Scientific
Hydroxyectoin 95%
Catalog # APOSOR1025890-5G
Supplier:  Apollo Scientific
New Product
CAS Number:  165542-15-4
Hydroxyectoin 95%
Bottle
CAS Number:  165542-15-4
Price
341,00 €
Log in for availability
1 * 5 g
Product Details & Documents
Usage: Organic building block/reagent.
  • CAS Number:165542-15-4
  • Formula:C₆H₁₀N₂O₃
  • Molecular Weight:158,16 g/mol
Specifications
Specifications
  • Pack type:
    Bottle
Specifications