Order Entry
Slovenia
Orders LinkContactUsLinkComponent
2-Chlorocyclohexanone 95%

2-Chlorocyclohexanone 95%

Bottle
Catalog # APOSOR918717-100G
New Product
Supplier:   Apollo Scientific
2-Chlorocyclohexanone 95%
Catalog # APOSOR918717-100G
Supplier:  Apollo Scientific
New Product
CAS Number:  822-87-7
2-Chlorocyclohexanone 95%

Bottle

CAS Number:  822-87-7
Price
62,30 €
Log in for availability
1 * 100 g
Product Details & Documents
  • CAS Number:822-87-7
  • Formula:C₆H₉ClO
  • MDL Number:MFCD00001626
  • Molecular Weight:132.59 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle

Specifications