ContactUsLinkComponent
1 component: S 50mg/l in Base oil 75 cSt CRM, ISO 17034 and ISO 17025 Conservation 12 months. Store under normal laboratory conditions, at temperatures between 15°C to 25°C 1 * 100 mL
Catalog # CMPLRM122375L1
New Product
1 component: S 50mg/l in Base oil 75 cSt CRM, ISO 17034 and ISO 17025 Conservation 12 months. Store under normal laboratory conditions, at temperatures between 15°C to 25°C 1 * 100 mL
Catalog # CMPLRM122375L1