Picloram
Picloram
EHERC16200000
- SMILES: Nc1c(Cl)c(Cl)nc(C(=O)O)c1Cl
- Inchi: InChI=1S/C6H3Cl3N2O2/c7-1-3(10)2(8)5(9)11-4(1)6(12)13/h(H2,10,11)(H,12,13)
- IUPAC: 4-amino-3,5,6-trichloropyridine-2-carboxylic acid
- Packing group: III
- Description:Picloram
- UN No.:3077
- Cat. No.:EHERC16200000
- Formula:C₆H₃Cl₃N₂O₂
- Storage conditions:20 °C
- Hazard class description:GHS07, GHS08, GHS09
- GHS signal:Warning
- Synonym(s):2-Pyridinecarboxylic acid, 4-amino-3,5,6-trichloro-;Picolinic acid, 4-amino-3,5,6-trichloro- (8CI);4-Amino-3,5,6-trichloro-2-pyridinecarboxylic acid;3,5,6-Trichloro-4-aminopicolinic acid;4-Amino-3,5,6-trichloro-2-picolinic acid;4-Amino-3,5,6-trichloropicolinic acid;4-Amino-3,5,6-trichloropyridine-2-carboxylic acid;4-Aminotrichloropicolinic acid;ATCP;Grazon PC;NSC 233899;Padron;Padron (herbicide);Picloram;Tordon
- Molecular weight:241,46
- CAS No.:1918-02-1
- Shipping conditions:RT



