n-Pentane
n-Pentane
Catalog # EHERC15977500
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CCCCC
- Inchi: InChI=1S/C5H12/c1-3-5-4-2/h3-5H2,1-2H3
- IUPAC: pentane
- Packing group: II
Specifications
Specifications
- Description:n-Pentane
- UN No.:1265
- Cat. No.:EHERC15977500
- Formula:C₅H₁₂
- Storage conditions:20 °C
- Hazard class description:GHS02, GHS07, GHS08, GHS09
- GHS signal:Danger
- Synonym(s):Pentane;E 0121;NSC 72415;Norpar 5S;Skellysolve A;n-Pentane
- Molecular weight:72,15
- CAS No.:109-66-0
- Shipping conditions:RT



