Tebufenozide
Tebufenozide
Catalog # EHERC17178800
Supplier: EHRENSTORFER
Jump to:
Product Details & Documents
- SMILES: CCc1ccc(cc1)C(=O)NN(C(=O)c2cc(C)cc(C)c2)C(C)(C)C Inchi: InChI=1S/C22H28N2O2/c1-7-17-8-10-18(11-9-17)20(25)23-24(22(4,5)6)21(26)19-13-15(2)12-16(3)14-19/h8-14H,7H2,1-6H3,(H,23,25) IUPAC: N-tert-butyl-N'-(4-ethylbenzoyl)-3,5-dimethylbenzohydrazide
- Packing group: III
Specifications
Specifications
- Description:Tebufenozide
- UN No.:3077
- Cat. No.:EHERC17178800
- Formula:C₂₂H₂₈N₂O₂
- Storage conditions:20 ℃
- Hazard class description:GHS09
- GHS signal:Warning
- Synonym(s):Benzoic acid, 3,5-dimethyl-, 1-(1,1-dimethylethyl)-2-(4-ethylbenzoyl)hydrazide (9CI, ACI); 1-tert-Butyl-1-(3,5-dimethylbenzoyl)-2-(4-ethylbenzoyl)hydrazine; 3,5-Dimethylbenzoic acid N-tert-butyl-N-(4-ethylbenzoyl)hydrazide; Confirm; Mimic; Mimic 240LV; Mimic 700WP; RH 5992; Romdan; Tebufenozide
- Molecular weight:352,47
- CAS No.:112410-23-8
- Shipping conditions:RT



